Learn More
Cole-Parmer™ Polypropylene Manifold
Expand your flow and line capacity
Brand: Cole-Parmer™ 31522-29
This item is not returnable.
View return policy
Description
- Constructed of durable polypropylene.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
Polypropylene manifold; 10 outlets; 1/2 in. inlet NPT(F) | |
27.31 cm | |
0°C | |
3.81 cm | |
150 psi | |
99°C |
Polypropylene | |
32°F | |
1/2 in. | |
3.81 cm | |
210°F |