Learn More
Labcold™ Powder Coated Steel Pharmacy Refrigerators
Brand:  Labcold™ RPFR21043
 View more versions of this product
Product Code. 12928614
  • 158785.00 SEK / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Description
  • Microprocessor controlled temperature regulation for accuracy and stability
  • Temperature range +2°C to +8°C
  • Fan circulated interior
  • Stainless steel construction
  • Door locks fitted as standard
  • Digital temperature display with audio alarm
  • Heavy duty lockable castors on larger models
  • Optimised air circulation
Specifications
Specifications
820 x 720 x 1980mm
Documents
One moment while we fetch your results.
Product Title
Labcold™ Powder Coated Steel Pharmacy Refrigerators > Exterior Dimensions (H x W x D): 1980 x 720 x 820mm