Learn More
Hanna Instruments™ Free Chlorine Reagents (100 tests)
High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements.
Brand:  Hanna Instruments™ HI9370101
Product Code. 10294031
  • 795.00 SEK / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Reagents for the colorimetric determination of free chlorine
  • There are enough reagents for 100 tests and are for use with Hanna’s compatible benchtop and portable photometers
  • High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
  • Follow the DPD Method in which the reaction between chlorine and reagent causes a pink tint in the sample 
  • Designed to be used with samples that have an expected range of 0.00 to 5.00 mg/L free chlorine
Specifications
Specifications
100 Pack