missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Thermo Scientific™ Precipitation Solution
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Kvantitet:
40 ml
missing translation for 'unitSize'
40ml
Description
Thermo Scientific GeneJET Precipitation Solution is a component of the GeneJET Plant RNA Purification Mini Kit (K0791/K0792) and may be purchased separately. Provided in 40 mL aliquots.
Specifications
Specifications
| Buffert | Precipitation Solution |
| Beskrivning | Precipitation Solution, 40mL, Liquid, Component of the GeneJET Plant Genomic DNA Purification Kit |
| Formulering | Liquid |
| För användning med (applikation) | DNA extraction, DNA & RNA purification & analysis, RNA extraction, total RNA isolation, total RNA from plant cells |
| Produkttyp | Precipitation Solution |
| Kvantitet | 40 ml |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction