Läs mer
Uvex Preformed Mask silv-Air e 7317 FFP3
Handla allt Uvex Produkter
:
Filter Type: FFP3
Beskrivning
General Features
- Particle-filtering preformed mask with exhalation valve
- Color: white
Protective Features
- FFP3 respirator certified in accordance with EN 149:2001+A1:2009
- D symbol (mask has passed the dolomite dust test): breathe easy even at high levels of dust exposure
- R certification (re-usable): tested for re-usability and performance during second shift
Comfort Features
- Soft material edges for increased comfort
- Very soft, all-around sealing lip for optimal wearer comfort and secure fit
- No nose clip (metal-free)
- Four-point headband attachment for a comfortable, individual and secure fit
- Extra-wide textile headband for enhanced comfort
- Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
Specifikationer
Specifikationer
| Certifieringar/efterlevnad (Europa) | EN 149 (R marked, D marked) |
| Produkttyp | Preformed Mask |
| Med utandningsventil | Yes |
| Filtereffektivitet | FFP3 |
| Inkluderar | 360° exhalation valve, four-point head-securing strap, circumferential comfort sealing lip |