Learn More
Novus Biologicals™ PRICKLE3 Overexpression Lysate
Brand: Novus Biologicals™ NBP2-07661
This item is not returnable.
View return policy
Description
The PRICKLE3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_6141. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens PRICKLE3, mRNA. | |
Native |
For Research Use Only