missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567090
This item is not returnable.
View return policy
Description
Immunogen sequence: THLSSTEAFCV FYHLKSNPSV FLCQCSCYVA EDQQYQWLEK VFGSCPRKNM QITILTCRHV TDYKTSESTG SLPSPF Highest antigen sequence identity to the following orthologs - mouse 77%, rat 75%.
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG2. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.Specifications
PSMG1 | |
Polyclonal | |
Unconjugated | |
PSMG1 | |
AW552102; C21LRP; C21-LRP; Chromosome 21 leucine-rich protein; Down syndrome critical region gene 2; Down syndrome critical region homolog 2; Down syndrome critical region protein 2; Down syndrome critical region protein 2 homolog; Dscr2; leucine rich protein C21-LRP; LRPC21; PAC1; PAC-1; proteasome (prosome, macropain) assembly chaperone 1; proteasome assembling chaperone 1; proteasome assembly chaperone 1; Psmg1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95456 | |
PSMG1 | |
Recombinant Human PSMG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only