Learn More
Molecular Probes™ Qdot™ Incubation Buffer
Qdot™ Incubation Buffer
Brand:  Molecular Probes™ Q20001MP
1153.39 SEK valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.01050kg
Product Code. 10098972
  • 1730.00 SEK / Each
Estimated Shipment: 12-07-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
Qdot™ Incubation Buffer - 2% BSA in 50 mM borate buffer, pH 8.3, containing 0.05% sodium azide is dentical to the incubation buffer (component B) provided in the Qdot™ ITK™ Streptavidin Conjugate Kits.
Specifications
Specifications
30 mL
Room Temperature
Qdot™
Store in refrigerator (2–8°C).