missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RadEye™ G Series Personal Dose Rate Meters

Detect and measure even the smallest change in gamma and dose rate measurements with intrinsically safe Thermo Scientific™ RadEye™ G Series of personal radiation detectors.

Brand:  Thermo Scientific™ 425067670

 View more versions of this product

Product Code. 15504789

  • 33090.00 SEK / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Continuously monitor your radiation exposure in any nuclear or radiological emergency. The Thermo Scientific™ RadEye™ G Series of personal radiation detectors provide first responders with quick, reliable gamma and dose rate measurements. These light-weight, handheld Geiger counters detect and measure very low gamma energies; even the smallest change in radiation rates are displayed immediately, while coincidentally occurring fluctuations are suppressed. For hazmat teams investigating potentially explosive atmospheres, intrinsically safe RadEye G models meet the latest ATEX standards.

The RadEye G Ex Series of Personal Radiation Detectors comprises four versions of intrinsically safe handheld devices for gamma and dose rate measurements for operation in and around hazardous areas. 

  • Efficient and reliable dose and dose rate measurements.
  • Intrinsically safe according to ATEX standards.
  • Large, clear and backlit display for error free readings.
  • Rugged and reliable.
  • Lightweight, only 160g (5.6 oz.).
  • Low power technology.
Recommended for:
  • Emergency services
  • Fire brigades
  • Hazmat teams
  • Locations with risk of explosion
  • Refineries
  • Oil platforms
Specifications

Specifications

II 2G Ex ia IIB T4 IBExU10ATEX1096
RadEye GF-10-Ex with Orange Label
IP 65 according to EN 60529
2 AAA
5 µSv/h to 3 Sv/h [500 urem/h - 300 rem/h]
up to 10 Sv/h
0.18 kg
900 hr.
H*(10) up to 10 Sv
Cs-137, 660 keV, ˜0.13cps/μSv/h
3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Personal Radiation Detector
Energy-compensated Geiger-Mueller tube
For Cs-137 (622 keV): 0.13 cps per μSv/h
H*(10) in Sv and Sv/h
0.41 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.