missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HPV B19 Capsid protein VP1 Protein
A DNA sequence encoding the Homo sapiens (Human) HPV B19 Capsid protein VP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7371-ec-200ug
This item is not returnable.
View return policy
Specifications
HPV B19 Capsid protein VP1 Protein | |
Research Use Only | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
200 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
MTSVNSAEAS TGAGGGGSNS VKSMWSEGAT FSANSVTCTF SRQFLIPYDP EHHYKVFSPA ASSCHNASGK EAKVCTISPI MGYSTPWRYL DFNALNLFFS PLEFQHLIEN YGSIAPDALT VTISEIAVKD VTDKTGGGVQ VTDSTTGRLC MLVDHEYKYP YVLGQGQDTL APELPIWVYF PPQYAYLTVG DVNTQGISGD SKKLASEESA FYVLEHSSFQ LLGTGGTASM SYKFPPVPPE NLEGCSQHFY EMYNPLYGSR LGVPDTLGGD PKFRSLTHED HAIQPQNFMP GPLVNSVSTK EGDSSNTGAG KALTGLSTGT SQNTRISLRP GPVSQPYHHW DTDKYVTGIN AISHGQTTYG NAEDKEYQQG VGRFPNEKEQ LKQLQGLNMH TYFPNKGTQQ YTDQIERPLM VGSVWNRRAL HYESQLWSKI PNLDDSFKTQ FAALGGWGLH QPPPQIFLKI LPQSGPIGGI KSMGITTLVQ YAVGIMTVTM TFKLGPRKAT GRWNPQPGVY PPHAAGHLPY VLYDPTATDA KQHHRHGYEK PEELWTAKSR VHPL | |
Greater than 90% as determined by SDS-PAGE. |