missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Splicing factor U2AF 26 kDa subunit Protein

A DNA sequence encoding the Homo sapiens (Human) Splicing factor U2AF 26 kDa subunit, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7888-ec-1mg

 View more versions of this product

Product Code. 16071411

  • 38570.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001035515.1
Splicing factor U2AF 26 kDa subunit Protein
Research Use Only
U2AF1L4
Recombinant Protein
E. coli
MAEYLASIFG TEKDKVNCSF YFKIGVCRHG DRCSRLHNKP TFSQEVFTEL QEKYGEIEEM NVCDNLGDHL VGNVYVKFRR EEDGERAVAE LSNRWFNGQA VHGNVPEVAS ATSCICGPFP RTSRGSSMGG DPGAGHPRGS ILATIPERGT IGVPLITGMA ASEALAPLPF TPNRDRCSWQ DLSSKPPSLS CPILPRLPGS IM
Greater than 80% as determined by SDS-PAGE.
199746
1 mg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers