missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
RhoJ Antibody (OTI1D7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45590
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
RhoJ Monoclonal specifically detects RhoJ in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifikationer
| RhoJ | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| ARHJTC10B, FLJ14445, ras homolog gene family, member J, RASL7B, Ras-like protein family member 7B, RAS-like, family 7, member B, RHOI, rho-related GTP-binding protein RhoJ, Tc10-like GTP-binding protein, TC10-like Rho GTPase, TCLMGC34777 | |
| Mouse | |
| 23.6 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 57381 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1D7 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| RHOJ | |
| Full length human recombinant protein of human RHOJ(NP_065714) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering