Learn More
Ansell Ringers 065 R-Flex Impact Glove
Click to view available options
Size:
7
8
9
10
11
12
13
Description
- The Ringers 065 R-Flex Impact glove with a Nitrile Half Dip palm combines protection against impact hazards with cut resistance for a wide range of applications.
- A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design provide superior dexterity and comfort.
- The sandy finish palm ensures secure grip in wet and dry environments, and touchscreen compatible index, middle and thumb tips allow technology interaction.
Specifications
Specifications
Coating Material | Nitrile |
Color | Hi-vis Yellow, Gray |
Cut Resistance Rating | ANSI A4 |
Hand | Specific |
Latex Free | Yes |
Material | HPPE |
Size | 7 |
Glove Cuff Style | Knit |