Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
Vapor duct with Combi-Clip for use with connecting the evaporating flask to the Rotavapor.
2440.00 SEK
Product Code | Brand | For Use With (Application) | Standard Taper Joint Number Center | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Standard Taper Joint Number Center | Price | Quantity & Availability | ||||
15475875
|
BUCHI
11062186 |
Glass Assembly V, C, S, E, CR, BY, HP | 29/32 |
2440.00 SEK
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
Description
- Combi-Clip is included for fastening the evaporating flask
Specifications
Borosilicate Glass 3.3 | |
Vapor Duct | |
Combi-Clip |