Learn More
3M™ S-Series Hood with Inner Shroud, Premium Head Suspension
S-Series hood with sealed seams features a double shroud design for its respiratory seal.
12900.00 SEK
Specifications
Assigned Protection Factor (APF) | 1000 |
---|---|
Material | Zytron 200 |
For Use With (Application) | Assembly, facility maintenance, machine operation, painting, sanding |
For Use With (Equipment) | GVP, Breathe-Easy™, V Series, T-300, T-600, T-800 |
Suspension Type | Premium Resusable Suspension |
Description
Premium suspension hoods and head covers
- Head covers provide head and face coverage; hoods cover head, face, and neck
- Fully assembled and ready to use
- Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or situations where there is less soiling of the outer fabric
- Integrated models feature wide face seal with soft elastic for improved comfort and reduced noise
Premium Reusable Suspensions with Hoods
- Ideal for high disposal rate environments
- Soiled hood is replaced; suspension and air ducting are reused
- Rigid ducting for consistent air path
- Externally adjustable airflow control allows wearer to modify airflow distribution
- Suspension features independent adjustments for the crown and head circumference; allows custom fit for wide range of head sizes
- 3M Catalog# S-805-5, S-807-5, S-855, S-857
Specifications
1000 | |
Assembly, facility maintenance, machine operation, painting, sanding | |
Premium Resusable Suspension |
Zytron 200 | |
GVP, Breathe-Easy™, V Series, T-300, T-600, T-800 | |
Acetate |