Learn More
WTW™ Sample Bottle SB VT 900
Sample bottle for use with the WTW™ IDS turbidity sensor.
Brand:  WTW™ 600704
Product Code. 16418743
  • 545.00 SEK / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Description
  • Sample bottle for the VisoTurb™ turbidity sensor
  • Accesssory for the IDS MultiLine™ meters
  • Capacity 500mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VisoTurb Turbidity Sensor
Sample bottle, 500 mL