Läs mer
BD Vacutainer Sharps Container
Kapacitet (metrisk):
0.7 L
Förpackningsstorlek:
Paket med 50
Beskrivning
- Available in a range of sizes and shapes.
- Designed for point-of-use disposal to ensure maximum safety.
- Provides an environment and an approach that is focused on safety to help reduce the risks of blood exposure and needle stick injuries associated with the disposal process.
- Compatible brackets for placement on flat surfaces, secured to walls, or for placement on medication carts.
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 0.7 L |
| Mått (L x B x H) | 89 x 74 x 168 mm |
| Kvantitet | 50/Pk. |