missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Sodium Free Salt Mixture

Sodium free salt mixture.

Brand:  MP Biomedicals 0290291002

 View more versions of this product

Product Code. 11445682

This item is not returnable. View return policy

Description

Description

  • Sodium free salt mixture
  • Contains calcium carbonate and potassium phosphate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Single-use
Non-sterile
1 ft.
Animal Diet
2 kg
Room Temperature
0.3048 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.