Läs mer
Labcold™ Sparkfree Fridge Freezer
Produktkod. 11528974
Modell:
Kylskåp/frys
Kylskåp/frys med nyckel
Förpackningsstorlek:
Varje
Produktkod. 11528974
Brand: Labcold™ RLFF13246/LK
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Stores ­flammable materials safely with no sources of ignition inside the chambers. Labcold™ Sparkfree Fridge Freezer provides all the routine storage required on a floor area no bigger than a standard fridge. For laboratory and general use where there is a concern that volatile, flammable materials or components might be stored now, or at some point in the future.
Combination sparkfree laboratory refrigerator and freezer provides all the routine storage required on a floor area no bigger than a standard fridge.
  • For laboratory and general use where there is a concern that volatile, flammable materials or components might be stored now, or at some point in the future.
  • Both refrigerated and frozen samples can be stored and accessed within the same small floor area.
  • Automatic defrosting in fridge
  • Drawers in Freezer are CFC and HCFC free
  • Two refrigeration systems - one for the freezer the other for the fridge
Specifikationer
Typ Spark-free Refrigerator Freezer
Kapacitet (metrisk) Frys 134 L
Kapacitet (metrisk) Kylskåp 228 L
Temperaturområde (frys) -18°C to -23°C
Temperaturområde (kylskåp) 2°C to 10°C
Antal lådor 4 (Freezer)
Hyllor 4 (Refrigerator)
Kylmedel Eco-friendly
Exteriör finish White Enamel
Interiör Molded Plastic
Inkluderar Sparkfree interior, 4 drawers in freezer, 4 shelves in fridge, and door lock with key
Modell Kylskåp/frys med nyckel
Djup (metrisk) Exteriör 630 mm
Höjd (metrisk) Exteriör 1965 mm
Bredd (metrisk) Exteriör 595 mm
Energianvändning 1.63kWh/24 hr.
Vikt (metrisk) 80.43 kg