Learn More
Hoefer™ Spring Clamps for Mighty Small™ Electrophoresis Units
For use with Mighty Small electrophoresis units
803.64 SEK - 1004.55 SEK
Specifications
Color Red
Type Spring Clamp
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10608105
View Documents Promotion Details
Hoefer™
SE252
SE 250, SE 260 Mini Vertical Electrophoresis Unit
1004.55 SEK
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
12805856
View Documents Promotion Details
Hoefer™
SE253
SE 250, SE 260 Mini Vertical Electrophoresis Unit, SE 615, SE 675 Multiple Gel Casters
803.64 SEK
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Multiple Gel Casters Replacement Parts
  • Some are Asymetrical Clamps
Specifications
Specifications
Red
Spring Clamp