Learn More
Agencinox™ Stainless Steel Footstool
Use this footstool to safely retrieve items from high places. Agencinox™ Stainless Steel Footstools feature non-slip steps and are designed for use in laboratories and medical environments.
2210.10 SEK - 5045.60 SEK
Specifications
Material | Stainless Steel |
---|---|
Product Type | Foot Stool |
Quantity | 1 Each |
Width (Metric) | 340 mm |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11709885
|
Agencinox™
759 |
210 mm |
2210.10 SEK
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
11744926
|
Agencinox™
779 |
180 mm |
5045.60 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Anti-skidding steps
- 18/10 Stainless steel construction
- Available in a variety of sizes
Specifications
Stainless Steel | |
Foot Stool | |
1 Each | |
340 mm |