Learn More
Fisherbrand™ Stainless Steel Manual Decapper for clean room
Allow easy removal of aluminum seals without breakage
9715.00 SEK - 13040.00 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11520535
|
Fisherbrand™
|
20mm Decapper |
9715.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
15259968
|
Fisherbrand™
|
32mm Decapper |
13040.00 SEK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11586034
|
Fisherbrand™
|
13mm Decapper |
9715.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
11845722
|
Fisherbrand™
|
11mm Decapper |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Stainless Steel
- Sterilizable for cleanroom applications
- Manual
- Autoclavable