Learn More
RETSCH Stainless-Steel Lid for Test Sieves
Cover sieves to avoid loss of sample during separation process. These stainless steel lids will fit multiple test sieves, based upon the sieve's diameter.
Brand:  RETSCH 60.107.000315
 View more versions of this product
Product Code. 11998124
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Stainless steel
  • Available for a variety of test sieves, based upon diameter
Specifications
Specifications
1 Ea.
ISO 3310 and ASTM E 11
315 mm
Sieve Lid
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RETSCH Stainless-Steel Lid for Test Sieves > Diameter: 315 mm