Learn More
Day Impex™ Stainless Steel Tempering Beaker
Click to view available options
Capacity (Metric):
600 mL
Description
- Double walled beaker
- Includes inlet and outlet ports for controlling temperature
- Constructed of stainless steel
Specifications
Specifications
Capacity (Metric) | 600 mL |
Material | Stainless Steel |
Type | Beaker |