Learn More
RETSCH Stainless-Steel Test Sieves, 100 mm Dia., in Micrometer Pore Sizes with ASTM E 11
Specifications
Quantity | 1 Each |
---|---|
Type | Test Sieve |
Certifications/Compliance | ASTM E 11 |
For Use With (Equipment) | Shaker AS 200 |
Includes | Certificate indicating Compliance |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10545252
|
RETSCH
60.108.000106 |
106 μm |
N/A
|
N/A | |||||
10369701
|
RETSCH
60.108.000125 |
125 μm |
N/A
|
N/A | |||||
10238681
|
RETSCH
60.108.000180 |
180 μm |
N/A
|
N/A | |||||
10731423
|
RETSCH
60.108.000020 |
20 μm |
N/A
|
N/A | |||||
10186541
|
RETSCH
60.108.000150 |
150 μm |
N/A
|
N/A | |||||
10739312
|
RETSCH
60.108.000212 |
212 μm |
N/A
|
N/A | |||||
10460032
|
RETSCH
60.108.000250 |
250 μm |
N/A
|
N/A | |||||
10319561
|
RETSCH
60.108.000025 |
25 μm |
N/A
|
N/A | |||||
10490032
|
RETSCH
60.108.000300 |
300 μm |
N/A
|
N/A | |||||
10400042
|
RETSCH
60.108.000355 |
355 μm |
N/A
|
N/A | |||||
10411962
|
RETSCH
60.108.000032 |
32 μm |
N/A
|
N/A | |||||
10399601
|
RETSCH
60.108.000038 |
38 μm |
N/A
|
N/A | |||||
10054891
|
RETSCH
60.108.000425 |
425 μm |
N/A
|
N/A | |||||
10379891
|
RETSCH
60.108.000500 |
500 μm |
N/A
|
N/A | |||||
10771803
|
RETSCH
60.108.000045 |
45 μm |
N/A
|
N/A | |||||
10094861
|
RETSCH
60.108.000053 |
53 μm |
N/A
|
N/A | |||||
10620072
|
RETSCH
60.108.000600 |
600 μm |
N/A
|
N/A | |||||
10166401
|
RETSCH
60.108.000063 |
63 μm |
N/A
|
N/A | |||||
10630072
|
RETSCH
60.108.000710 |
710 μm |
N/A
|
N/A | |||||
10526022
|
RETSCH
60.108.000850 |
850 μm |
N/A
|
N/A | |||||
10535252
|
RETSCH
60.108.000075 |
75 μm |
N/A
|
N/A | |||||
10106501
|
RETSCH
60.108.000090 |
90 μm |
N/A
|
N/A | |||||
Specifications
1 Each | |
ASTM E 11 | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Test Sieve | |
Shaker AS 200 | |
40mm | |
100 mm | |
Stainless Steel |