Learn More
RETSCH Stainless-Steel Test Sieves, 450 mm Dia., in Millimeter Pore Sizes with ASTM E 11
Specifications
Quantity | 1 Each |
---|---|
Type | Test Sieve |
Certifications/Compliance | ASTM E 11 |
For Use With (Equipment) | Vibratory Shaker AS 450 |
Includes | Certificate indicating Compliance |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11944712
|
RETSCH
60.169.001700 |
1.7 mm |
N/A
|
N/A | |||||
11914712
|
RETSCH
60.169.001180 |
1.18 mm |
N/A
|
N/A | |||||
11924742
|
RETSCH
60.169.106000 |
106 mm |
N/A
|
N/A | |||||
11914742
|
RETSCH
60.169.100000 |
100 mm |
N/A
|
N/A | |||||
11954722
|
RETSCH
60.169.011200 |
11.2 mm |
N/A
|
N/A | |||||
11924712
|
RETSCH
60.169.001400 |
1.4 mm |
N/A
|
N/A | |||||
11964722
|
RETSCH
60.169.012500 |
12.5 mm |
N/A
|
N/A | |||||
11934742
|
RETSCH
60.169.125000 |
125 mm |
N/A
|
N/A | |||||
11984722
|
RETSCH
60.169.016000 |
16 mm |
N/A
|
N/A | |||||
11994722
|
RETSCH
60.169.019000 |
19 mm |
N/A
|
N/A | |||||
11974722
|
RETSCH
60.169.013200 |
13.2 mm |
N/A
|
N/A | |||||
11974712
|
RETSCH
60.169.002800 |
2.8 mm |
N/A
|
N/A | |||||
11904712
|
RETSCH
60.169.001000 |
1 mm |
N/A
|
N/A | |||||
11954712
|
RETSCH
60.169.002360 |
2.36 mm |
N/A
|
N/A | |||||
11904732
|
RETSCH
60.169.022400 |
22.4 mm |
N/A
|
N/A | |||||
11914732
|
RETSCH
60.169.025000 |
25 mm |
N/A
|
N/A | |||||
11924732
|
RETSCH
60.169.026500 |
26.5 mm |
N/A
|
N/A | |||||
11934712
|
RETSCH
60.169.002000 |
2 mm |
N/A
|
N/A | |||||
11964712
|
RETSCH
60.169.003350 |
3.35 mm |
N/A
|
N/A | |||||
11934732
|
RETSCH
60.169.031500 |
31.5 mm |
N/A
|
N/A | |||||
11954732
|
RETSCH
60.169.037500 |
37.5 mm |
N/A
|
N/A | |||||
11984712
|
RETSCH
60.169.004750 |
4.75 mm |
N/A
|
N/A | |||||
11944732
|
RETSCH
60.169.045000 |
45 mm |
N/A
|
N/A | |||||
11994712
|
RETSCH
60.169.004000 |
4 mm |
N/A
|
N/A | |||||
11904722
|
RETSCH
60.169.005600 |
5.6 mm |
N/A
|
N/A | |||||
11974732
|
RETSCH
60.169.053000 |
53 mm |
N/A
|
N/A | |||||
11914722
|
RETSCH
60.169.006300 |
6.3 mm |
N/A
|
N/A | |||||
11964732
|
RETSCH
60.169.050000 |
50 mm |
N/A
|
N/A | |||||
11924722
|
RETSCH
60.169.006700 |
6.7 mm |
N/A
|
N/A | |||||
11984732
|
RETSCH
60.169.063000 |
63 mm |
N/A
|
N/A | |||||
11994732
|
RETSCH
60.169.075000 |
75 mm |
N/A
|
N/A | |||||
11944722
|
RETSCH
60.169.008000 |
8 mm |
N/A
|
N/A | |||||
11934722
|
RETSCH
60.169.009500 |
9.5 mm |
N/A
|
N/A | |||||
11904742
|
RETSCH
60.169.090000 |
90 mm |
N/A
|
N/A | |||||
Specifications
1 Each | |
ASTM E 11 | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Test Sieve | |
Vibratory Shaker AS 450 | |
100mm | |
450 mm | |
Stainless Steel |