Läs mer
Azpack™ Unex™ Tubing Clips
Diameter (metrisk):
10 to 14 mm
13 to 17 mm
14 to 19 mm
18 to 22 mm
21 to 25 mm
26 to 30 mm
28 to 33 mm
8 to 10 mm
Förpackningsstorlek:
Paket med 5
Beskrivning
- Stainless steel
- Double-banded ring for even pressure
- Nut and bolt included to apply proper compression
Specifikationer
Specifikationer
| Typ | Tubing Clamp |
| Material | Stainless Steel (H10 304, A2) |
| Inkluderar | Tightening bolt |
| Diameter (metrisk) | 18 to 22 mm |
| Typ av käke | Double-banded Ring |