Läs mer
Azpack™ Unex™ Tubing Clips
Produktkod. 11398093
Diameter (metrisk):
10 till 14 mm
13 till 17 mm
14 till 19 mm
18 till 22 mm
21 till 25 mm
26 till 30 mm
28 till 33 mm
8 till 10 mm
Förpackningsstorlek:
Paket med 5
Produktkod. 11398093
Brand: Azpack™ 11398093
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Twist nut to constrict and release tubing. Azpack™ Stainless Steel Tubing Clips are designed to distribute and apply pressure evenly.
  • Stainless steel
  • Double-banded ring for even pressure
  • Nut and bolt included to apply proper compression
Specifikationer
Typ Tubing Clamp
Material Stainless Steel (H10 304, A2)
Inkluderar Tightening bolt
Diameter (metrisk) 8 till 10 mm
Typ av käke Double-banded Ring