Learn More
Usbeck Laborgeraete™ Steel/Ceramic Wire Cloths
78.10 SEK - 102.74 SEK
Specifications
Ceramic Center | Ceramic Center |
---|---|
Material | Steel, Ceramic |
Type | Wire Cloth |
Edges | Folded |
Quantity | 1 Each |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10372901
|
Usbeck Laborgeraete™
5081 |
120 mm |
78.10 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
10732702
|
Usbeck Laborgeraete™
5082 |
160 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Ceramic Center | |
Wire Cloth | |
1 Each |
Steel, Ceramic | |
Folded | |
Square |