Learn More
R&L Enterprises™ Steel Gas Cylinder Stands
Provides sturdy, secure support for gas cylinders in the laboratory. R&L Enterprises™ Steel Gas Cylinder Stands are strongly constructed in nylon-coated steel.
2030.00 SEK - 2185.00 SEK
Specifications
Material | Steel |
---|---|
Product Type | Gas Cylinder Stand |
For Use With (Equipment) | Cylinder |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12975961
|
R&L Enterprises™
3604 |
146 mm dia. cylinder |
2030.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12985961
|
R&L Enterprises™
3605 |
178 mm dia. cylinder |
2185.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Available in two different sizes
- Dimension given is the maximum diameter of cylinder accomodated
Specifications
Steel | |
Gas Cylinder Stand | |
Cylinder |