Läs mer
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Produktkod. 12806466 Handla allt IBA Lifesciences Produkter
Kvantitet:
1 mL
5 mL
5 x 0.2 mL
Förpackningsstorlek:
1ml
Paket med 5
Produktkod. 12806466
Brand: IBA Lifesciences 21209001
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Gravity flow kolonn för att rena Strep-™tag II eller Twin-Strep-tag™ fusionsproteiner
  • Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifikationer
Kvantitet 1 mL
Endast för forskning.