Läs mer
Systec™ Systec™ Air Exhaust Filtration Cartridge for Systec™ H-210 to H-1580
Produktkod. 12552777
För användning med (utrustning):
Systec H-210 to H-1580 Autoclaves
Förpackningsstorlek:
Varje
Produktkod. 12552777
Brand: Systec™ 8032
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Systec™ Air Exhaust Filtration Cartridge for Systec™ H-210 to H-1580 filters autoclave exhaust to prevent the escape of hazardous biologicals during sterilization.
  • For the sterilization of hazardous biological substances, Systec™ autoclaves can be fitted with an optional air exhaust filtration system.
  • The autoclavable sterile filter, consisting of a filter cartridge with PTFE membrane of pore size 0.2 μm, is installed in a pressure-proof housing and can be quickly changed at any time.
  • The filter is also automatically sterilized inline during the sterilization process, monitored by the PT-100 temperature sensor.
Specifikationer
Design 0.2μm pore size
För användning med (utrustning) Systec H-210 to H-1580 Autoclaves
Material PTFE