Läs mer
Hanna Instruments™ TDS Tester Primo 2
Produktkod. 11533729
Mått (L x B x H):
180 x 50 x 25 mm
Förpackningsstorlek:
Varje
Produktkod. 11533729
Brand: Hanna Instruments™ PRIMO 2
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
  • TDS/conductivity measurement
  • Low battery LCD indicator
  • Automatic temperature compensation (ATC)
  • Auto-shut off
  • Auto-calibration
Specifikationer
Kalibrering Automatic, at 1382 ppm
Temperaturkompensation Automatic, 0 to 60°C (32 to 122°F); β = 2%/°C
Noggrannhet (temperatur) At 20°C
Mått (L x B x H) 180 x 50 x 25 mm
Vikt (metrisk) 50 g
Garanti 1 Year
Sondtyp Direct, fixed
Upplösning 1 ppm (mg/L); 0.1°C
Typ TDS Tester Primo 2
Noggrannhet ±2 % F.S. / ±0.5.°C
Batteritid Approximately 200 hr., auto-off after 5 min. of non-use
Spänning 2 x 1.5 V