Learn More
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™
895.40 SEK - 8363.88 SEK
Chemical Identifiers
CAS 864771-44-8
Molecular Formula C18H25BN2O4
Molecular Weight (g/mol) 344.22
MDL Number MFCD08437629
InChI Key MMXDKXUKSKMROG-UHFFFAOYSA-N
Synonym tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-boc-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-n-boc-indazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-indazole-1-carboxylic acid tert-butyl ester, 1-boc-indazole-5-boronic acid pinacol ester, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1h-indazole-1-carboxylic acid, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate
PubChem CID 18525862
IUPAC Name tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1N=CC2=CC(=CC=C12)B1OC(C)(C)C(C)(C)O1
View More Specs
Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10357584
SDS View Documents
Thermo Scientific Maybridge
CC53239CB
250 mg Amber Glass Bottle
895.40 SEK
250mg
10366843
SDS View Documents
Thermo Scientific Maybridge
CC53239DA
1 g Amber Glass Bottle
3018.71 SEK
1g
11356835
SDS View Documents
Thermo Scientific Maybridge
CC53239DE
5 g Amber Glass Bottle
8363.88 SEK
5g
Specifications
Show More
Videos
Safety and Handling
Safety and Handling
Warning
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™