missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Phosphate, total LR Tube Test, 1.5 to 20mg/L and 5 to 60mg/L
Use these tests with test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 2420700
This item is not returnable.
View return policy
Description
Use these tests with test kits, colorimeters and spectrophotometers
- Parameter: Total Phosphate
- Method: Phosphomolybdic acid
- For use in test kits, colorimeters and spectrophotometers
- Case of 24
- Includes:
- Reaction Tube, LR
- Phosphate-101
- 2 Calibration Tubes
- Phosphate-102 Solution
- Phosphate-103 Reagent
Specifications
Test Kits, Colorimeters and Spectrophotometers | |
24 | |
Phosphomolybdic acid |
Reaction Tube, LR, Phosphate-101, Calibration Tube - 2 ct., Phosphate-102 Solution, Phosphate-103 Reagent | |
1.5 to 20mg/L and 5 to 60mg/L | |
Phosphate, Total LR Tube Test |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- total phosphate HR
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : sulphuric aicd
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction