Läs mer
Tris SDS PAGE Running Buffer, FASTRun, 10X
Kvantitet:
500mL
1L
Förpackningsstorlek:
1 L
500ml
Beskrivning
- Cut the time you spend waiting for your SDS PAGE gels to finish by over 70% while providing comparable or better clarity over traditional buffers
- Novel buffer chemistry gives you better resolution, thus requiring fewer TG gel compositions to fully resolve a protein.
Specifikationer
Specifikationer
| CAS | 151-21-3 |
| Färg | Colorless to Yellow |
| pH-intervall | 7.7 to 8.3 (1X/10X soln. at 25°C) |
| Form | Liquid |
| Kvantitet | 1L |