missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
TSLPR/CRLF2 Antibody (59N5G4), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27331AF405
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TSLPR/CRLF2 Monoclonal specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| TSLPR/CRLF2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Q9HC73 | |
| CRLF2 | |
| Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Immunology | |
| 64109 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 59N5G4 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| TSLPR is a glycoprotein with two major isoforms of 359 amino acids and 255 amino acids. In western blotting, bands may be visualized between 27 kD and ∼50 kD. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering