Learn More
Tocris Bioscience™ UNC 1999
Potent and selective EZH2/EZH1 inhibitor
Brand: Tocris Bioscience™ 4904/10
2268.62 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Description
UNC 1999 is a potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.Negative Control also available.
Specifications
>99% | |
UNC 1999 | |
UNC 1999 |
10 mg | |
569.74 |
For Research Use Only