Läs mer
Watson-Marlow 323 Series Drive with 313DW Pumphead
Pumphuvud:
313D
323Du
323E/D
323S/D
323U/D
Förpackningsstorlek:
Varje
Beskrivning
- 300 series laboratory pumps are a benchtop pumping solution with manual, remote, analogue or RS232 control capabilities. Featuring a chemically resistant ABS top and IP31 protection, 323E/D pumps have a membrane keypad that controls the pump, and a digital LCD to display speed. There are four models with increasing degrees of functionality:
- 323E/DDu RS232/auto/manual enhanced-control high-flow pumps
- 323E/DU and 323E/DU analogue-auto and enhanced manual-control pumps
- 323E/DS high-performance manual control pumps
- 323E/DE manual control pumps
Pumphead options available as standard (others available on request):
- 313D and 313D2 three-roller pumpheads for continuous tubing
- 314D and 314D2 four-roller pumpheads for continuous tubing
- 314MC four-roller five-channel microcassette pumpheads
- 318MC eight-roller five-channel microcassette pumpheads
- 501RL and 501RL2 pumpheads for continuous tubing
Specifikationer
Specifikationer
| Kontrolltyp | Variabel hastighet |
| Visningstyp | LCD |
| Kapslingsklassificering | IP31 |
| Flödeshastighet | 4,1 till 2000 ml/min. |
| Motorhastighet | 15 to 400 rpm |
| Reversibel | Ja |
| Pumphuvud | 323E/D |
| Hastighetskontroll | Keypad, manual control |
| Mått (L x B x H) | 230 x 230 x 132 mm |
| Spänning | 100/120/220/240 V |