Läs mer
Thermo Scientific™ Virtuoso™ Vial Identification System
Produktkod. p-6570125
Förpackningsstorlek:
Varje
Produktkod. 15201918
Brand: Thermo Scientific™ 60180VT100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Eliminate errors from manual vial labeling. This advanced labeling-information device ensures accurate sample identification and increases productivity.
The Thermo Scientific™ Virtuoso™ Vial Identification System is an error-free option to manual vial labeling that turns a necessary chore into a value-added component within a laboratory workflow. With advanced labeling capabilities, the Virtuoso system ensures accurate vial identification and reliable data, and increases laboratory productivity. Use the system’s touch screen and network capabilities to transfer text, graphics, barcodes, and logos directly onto your vials.
Facilitates Your Labeling Requirements
  • Fully loaded with standard or fully user-defined templates
Increases Productivity
  • Eliminate error-prone manual vial labeling
  • Label a vial in just 5 seconds
  • Improve labeling efficiencies by downloading sample lists
  • Simplify vial sample identification
Produces High-Quality Labeling
  • Resists common laboratory solvents, blurring, and smudging
Provides an Advanced Labeling and information System in One Device
  • Designed by experienced analytical scientists
TRUSTED_SUSTAINABILITY
Specifikationer
Produktlinje Virtuoso
Produkttyp Vial Identification System
Certifikat och efterlevnad Pollution Level 2
Höjd (engelska) 11.61 in.
Höjd (metrisk) 295 mm
Längd (engelska) 7.87 in.
Längd (metrisk) 200 mm
Strömförsörjning Universal power supply 100-240V AC
Bredd (metrisk) 140 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.