missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ VSIG4 Monoclonal Antibody (JAV4), Super Bright™ 436, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 62-5757-42
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This JAV4 monoclonal antibody recognizes human V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. There are two variants of human VSIG4 that result from alternative sp
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Specifikationer
| VSIG4 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q9Y279 | |
| VSIG4 | |
| Affinity chromatography | |
| RUO | |
| 11326 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| JAV4 | |
| Super Bright 436 | |
| VSIG4 | |
| A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering