Läs mer
Palintest™ Water Testing Tablet
För användning med (applikation):
Kalciumhårdhet
Kloridjoner
Total alkalinitet
Förpackningsstorlek:
Paket med 250
Beskrivning
- Ideal for quick and reliable testing in the field
- They are crushed in the water sample until a color change occurs and the number required allows results to be calculated in ppm or mg/L
- Magnesium hardness is the difference between total and calcium hardness
- Permanent hardness is the difference between total hardness and total alkalinity
- In bottles of 250 tablets with instructions
Specifikationer
Specifikationer
| För användning med (applikation) | Total alkalinitet |
| Kvantitet | 250 Pack |
| Typ | Water Testing Tablet |