missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wax, Pure Beeswax, Fisher Chemical
Click to view available options
250g, Plastic powder jar
Specifications
CAS | 8012-89-3 |
Melting Point | 61°C |
Packaging | Plastic powder jar |
MDL Number | 132754 |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'