man

Test

CAS RN 100-55-0

HON

CAS RN 100-55-0

IUPAC Name: (pyridin-3-yl)methanol
Synonyms: pyridin-3-ylmethanol nicotinyltartrate 3-picolyl alcohol nicotinyl alcohol β-picolyl alcohol nicotinic alcohol radecol hydrofluoride, nicomethanol ronicol retard β pyridyl carbinol (substance)
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: MVQVNTPHUGQQHK-UHFFFAOYSA-N
SMILES: OCC1=CC=CN=C1
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)