accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1000279-69-5

CAS RN 1000279-69-5

IUPAC Name: 4-[5-(4-chlorophenyl)-2-methyl-3-propanoyl-1H-pyrrol-1-yl]benzene-1-sulfonamide
Synonyms: 4-[5-(4-chlorophenyl)-2-methyl-3-propanoylpyrrol-1-yl]benzenesulfonamide
Molecular Weight (g/mol): 402.89
Molecular Formula: C20H19ClN2O3S
InChi Key: ABACVOXFUHDKNZ-UHFFFAOYSA-N
SMILES: CCC(=O)C1=C(C)N(C(=C1)C1=CC=C(Cl)C=C1)C1=CC=C(C=C1)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products