man

Test

CAS RN 100047-36-7

NNH2OHO

CAS RN 100047-36-7

IUPAC Name: 4-aminopyridine-2-carboxylic acid
Synonyms: 4-aminopyridine-2-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: JRZBTJVSAANBEV-UHFFFAOYSA-N
SMILES: NC1=CC(=NC=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results