CAS RN 100165-88-6
CAS RN 100165-88-6
IUPAC Name:
{2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane
Synonyms:
{2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane
Molecular Weight (g/mol):
678.8
Molecular Formula:
C48H40P2
InChi Key:
IOPQYDKQISFMJI-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals
CAS: 100165-88-6 Molecular Formula: C48H40P2 Molecular Weight (g/mol): 678.796 MDL Number: MFCD00269856 InChI Key: IOPQYDKQISFMJI-UHFFFAOYSA-N Synonym: r-tol-binap,s-tol-binap,s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,s-t-binap,rac-tol-binap,r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,tol-binap,s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,s-p-tol-binap PubChem CID: 4189906 IUPAC Name: [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane SMILES: CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C