man

Test

CAS RN 10022-68-1

CdOHNOOOHNOOH2OH2OH2OH2O

CAS RN 10022-68-1

IUPAC Name: cadmium(2+) bis(nitric acid)
Synonyms: cadmium nitrate
Molecular Weight (g/mol): 238.44
Molecular Formula: CdH2N2O6
InChi Key: IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES: [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Grade 
  • (2)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)