man

Test

CAS RN 10031-27-3

BrTeBrBrBr

CAS RN 10031-27-3

IUPAC Name: tetrabromo-λ⁴-tellane
Synonyms: tellurium tetrabromide
Molecular Weight (g/mol): 447.22
Molecular Formula: Br4Te
InChi Key: PTYIPBNVDTYPIO-UHFFFAOYSA-N
SMILES: Br[Te](Br)(Br)Br
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)