man

Test

CAS RN 10039-26-6

OOOOHOHHOHOOHOHOHHOH2O

CAS RN 10039-26-6

IUPAC Name: 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
Synonyms: starch hydrate
Molecular Weight (g/mol): 360.31
Molecular Formula: C12H24O12
InChi Key: WSVLPVUVIUVCRA-UHFFFAOYNA-N
SMILES: O.OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results